Learn More
Cayman Chemical R- LInoleyl-2-Hydroxy-1 10mg
SDP
Supplier:  Cayman Chemical 900123410 MG
Encompass_Preferred
A homolog of LOEA, characterized by the addition of an (R)-α-methyl group at the methylene carbon adjacent to the amide nitrogen
Catalog No. 50-289-3986