Learn More
Medchemexpress LLC (R)-lisofylline | 100324-81-0 | MFCD00867573 | 100.0% | 280.32 g/mol | C13H20N4O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY109854A5MG
Encompass_Preferred
(R)-Lisofylline is the R-enantiomer of lisofylline, a metabolite of pentoxifylline with documented anti-inflammatory activity. It is used as a research compound to probe lipid signaling and immune pathways, including inhibition of lysophosphatidic acid acyltransferase and disruption of IL-12-mediated STAT4 activation.
  • R-enantiomer of lisofylline.
  • Lysophosphatidic acid acyltransferase inhibitor (reported IC50 ≈ 0.6 μM).
  • Modulates IL-12-STAT4 signaling relevant to immunology research.
  • Provided in small milligram research packs for laboratory use.
  • Store under nitrogen at -20°C; in solvent, -80°C for up to 6 months.
  • Molecular weight 280.32 g/mol; formula C13H20N4O3.
Catalog No. 50-002-63404