Learn More
TARGETMOL CHEMICALS INC R-LISOFYLLINE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T216945MG
Encompass_Preferred
Also available in 1 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (R)-Lisofylline ((R)-Lisophylline) is an inhibitor of lysophosphatidic acid acyltransferase (IC50 = 0.6 µM). (R)-Lisofylline interrupts IL-12 signaling-mediated STAT4 activation and can be used in studies about the treatment of type 1 diabetes and autoimmune disorders. purity: 99%
Catalog No. 50-258-1011