Learn More
Medchemexpress LLC (R)-lunresertib ((R)-RP-6306) | 2719793-91-4 | 99.9% | C18H20N4O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY145817B10MM/1ML
Encompass_Preferred
(R)-lunresertib is an inhibitor of membrane-associated tyrosine and threonine-specific cdc2-inhibitory kinase (Myt1), also known by the gene name PKMYT1. It has an IC50 of 1360 nM and is utilized in the study of Myt1-mediated cancers. This product is an off-white to gray solid.
  • Inhibits Myt1 (PKMYT1) with an IC50 of 1360 nM
  • Used for research into Myt1-mediated cancers
  • Appears as an off-white to gray solid
  • Soluble in DMSO at 100 mg/mL
  • For research use only, not for patient use
Catalog No. 50-003-54013