Learn More
Medchemexpress LLC R-lunresertib ((R)-RP-6306) | 2719793-91-4 | 99.9% | C18H20N4O2 | 100 MG

Supplier: Medchemexpress LLC HY145817B100MG
(R)-lunresertib is a membrane-associated tyrosine and threonine-specific cdc2-inhibitory kinase (Myt1) inhibitor, with an IC50 of 1360 nM. It can be used in the study of Myt1-mediated cancers.
- Myt1 (PKMYT1) inhibitor
- Used in the study of Myt1-mediated cancers
- Research use only
- Appearance: Solid
- Color: Off-white to gray
- Molecular weight: 324.38
- Soluble in DMSO at 100 mg/mL
- Target: Wee1
- Pathway: Cell cycle/DNA damage
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.