Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC R-mephenytoin | 71140-51-7 | 99.6% | 218.25 g/mol | C12H14N2O2 | 100MG
SDP

Supplier:  Medchemexpress LLC HY126043100MG

Encompass_Preferred

(R)-Mephenytoin is the R-enantiomer of the anticonvulsant mephenytoin, provided as a research-grade solid for biochemical and pharmacological studies. It is commonly used in in vitro metabolism assays and cytochrome P450 interaction studies, notably to probe CYP2C9-mediated N-demethylation.

  • High purity (99.6%).
  • Molecular weight 218.25 g/mol.
  • Chemical formula C12H14N2O2.
  • Solid, white to off-white appearance.
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Supplied in small research quantities (e.g., 100 mg) for laboratory use.

Catalog No. 50-002-39136


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.