Learn More
Cayman Chemical R-Metalaxyl-d6 10mg
SDP
Supplier:  Cayman Chemical 3957510 MG
Encompass_Preferred
An internal standard for the quantification of (R)-(−)-metalaxyl by GC- or LC-MS
Catalog No. 50-291-8312