Learn More
Medchemexpress LLC (R)-N-(3-(4-(Methoxymethyl)-6-(3-methoxytetrahydrofuran-3-yl)pyridin-2-yl)-1-(oxetan-3-yl)-1H-pyrro... | 2368945-27-9 | 99.9% | 452.50 | C24H28N4O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15640910MG
Encompass_Preferred
ABBV-712 is a selective, orally active inhibitor of tyrosine kinase 2 (TYK2) developed as a clinical candidate for research into autoimmune and inflammatory diseases. It stabilizes the TYK2 pseudokinase (JH2) domain and demonstrates anti-inflammatory activity in preclinical models.
  • Selective TYK2 inhibition suitable for signaling and pharmacology studies.
  • Demonstrated anti-inflammatory activity in animal disease models.
  • High analytical purity for biochemical and cellular assays.
  • High solubility in DMSO (250 mg/mL) for concentrated stock solutions.
  • Available in small research pack sizes for dose-response testing.
  • Recommended storage: powder at -20°C for long term, 4°C for short term.
Catalog No. 50-002-87937