Learn More
Medchemexpress LLC (R)-(-)-N-benzyl-2-phenylglycinol | 14231-57-3 | MFCD00674035 | 98.0% | 227.31 g/mol | C15H17NO | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1418951G
Encompass_Preferred
(R)-(-)-N-Benzyl-2-phenylglycinol is a chiral amino-acid derivative intended for research and analytical applications. It functions as a chiral building block and intermediate in asymmetric synthesis and related synthetic chemistry. The compound is a white to off-white solid (CAS 14231-57-3) with formula C15H17NO and molecular weight 227.31 g/mol.
  • Chiral building block for asymmetric synthesis.
  • Intermediate for medicinal and synthetic chemistry applications.
  • Available in small-scale quantities for research (1 G and larger).
  • Solid form with storage recommendations to protect stability.
  • High assay purity suitable for research applications (assay reported at 98%).
Catalog No. 50-002-82364