Learn More
Medchemexpress LLC (R)-(-)-N-Benzyl-2-phenylglycinol | 14231-57-3 | MFCD00674035 | 98.0% | 227.31 g/mol | C15H17NO | 5 G

Supplier: Medchemexpress LLC HYW1418955G
(R)-(-)-N-Benzyl-2-phenylglycinol is the R-enantiomer of N-benzyl-2-phenylglycinol, a chiral amino-alcohol used as a research reagent and synthetic intermediate in organic and medicinal chemistry. Supplied as a solid for laboratory research, it should be handled and stored protected from light at low temperature.
- Chiral R-enantiomer reagent.
- Cas number 14231-57-3.
- Molecular formula C15H17NO and molecular weight 227.31 g/mol.
- Solid, white to off-white appearance.
- Store at 4°C and protect from light; in solution store at -20°C to -80°C depending on duration.
- Typical purity 98.0%.
- Available as a 5 G pack.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.