Learn More
STA PHARMACEUTICAL US LLC (R)-N-Fmoc-2-amino-oct-7-ynoic acid | 1 g | CAS 2350405-99-9 | InChIKey JGAAQTGBSLHOGR-OAQYLSRUSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAA102991GM
(R)-N-Fmoc-2-amino-oct-7-ynoic acid is a Amino Acid reagent (Subcategory: Azido/Alkynyl AA) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 2350405-99-9
- MDL: No data
- InChIKey: JGAAQTGBSLHOGR-OAQYLSRUSA-N
- Molecular Weight: 377.44
- Molecular Formula: C23H23NO4
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)oct-7-ynoic acid
- SMILES: C#CCCCC[C@@H](NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)C(O)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.