Learn More
TARGETMOL CHEMICALS INC R-Naproxen 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0855200MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. (R)-Naproxen, an anti-inflammatory agent, has antipyretic and analgesic properties. Both the acid and its sodium salt are used in the therapy of acute gout or dysmenorrhea, musculoskeletal disorders, rheumatoid arthritis and other rheumatic. Purity 99.2%
Catalog No. 50-288-0768