Learn More
Medchemexpress LLC (R)-Naproxen | 23979-41-1 | C14H14O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15031100MG
Encompass_Preferred
l-Naproxen ((R)-Naproxen) is an enantiomer of (S)-Naproxen. It can inhibit Cdc42 and Rac1 with EC50 values of 96 μM and 212 μM, respectively, and exhibits anti-tumor activity. l-Naproxen is also a nonsteroidal anti-inflammatory agent (NSAID). In HeLa cells, l-Naproxen selectively inhibits Rac1 and Cdc42 activity in a dose-dependent manner. It also inhibits the migration of immortalized human ovarian cancer cells and demonstrates activity through a COX-independent mechanism.
  • Inhibits Cdc42 and Rac1 with EC50 values of 96 μM and 212 μM
  • Demonstrates anti-tumor activity
  • Acts as a nonsteroidal anti-inflammatory agent (NSAID)
  • Selectively inhibits Rac1 and Cdc42 activity in HeLa cells
  • Inhibits migration of immortalized human ovarian cancer cells
  • Shows activity via a COX-independent mechanism
Catalog No. 50-003-75488