Learn More
Cayman Chemical R-nItro-BlebbIstatIn 10mg
SDP
Supplier:  Cayman Chemical 900193510 MG
Encompass_Preferred
A more stable form of (+)-blebbistatin, the inactive form of blebbistatin; has the same stereochemistry as the inactive (+)-blebbistatin enantiomer
Catalog No. 50-289-4478