Learn More
Medchemexpress LLC R-NVS-ZP7-4 | 100MG
SDP
Supplier:  Medchemexpress LLC HY114395A100MG
Encompass_Preferred
(R)-NVS-ZP7-4 is the R-isomer of NVS-ZP7-4 NVS-ZP7-4 is a Zinc transporter SLC39A7 (ZIP7) inhibitor that is also the first reported chemical tool to probe the impact of modulating ER zinc levels and investigate ZIP7 as a novel agentgable node in the Notch pathway
Catalog No. 50-002-29061