Learn More
Medchemexpress LLC R-NVS-ZP7-4 50mg | 2517682-14-1 | 50 MG

Supplier: Medchemexpress LLC HY114395A50MG
(R)-NVS-ZP7-4 is the R-enantiomer of NVS-ZP7-4, a small-molecule inhibitor of the zinc transporter SLC39A7 (ZIP7) supplied as an optically pure research compound for biochemical and cellular studies targeting ER zinc homeostasis, Notch signaling, and ferroptosis-related pathways.
- Optically pure R-enantiomer for stereochemistry-specific studies.
- High purity (~98.8%) suitable for biochemical and cellular assays.
- Solid, light yellow to yellow form; stable when stored at -20°C.
- Solvent storage: -80°C for up to 6 months, -20°C for up to 1 month (protect from light).
- Useful for probing ZIP7 function, ER zinc levels, Notch signaling, and ferroptosis mechanisms.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.