Learn More
Medchemexpress LLC (R)-OP-1074 | 1443752-77-9 | 98.2% | 457.56 g/mol | C29H31NO4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY125263B10MG
Encompass_Preferred
(R)-OP-1074 is the R-enantiomer of OP-1074, a selective estrogen receptor degrader used as a research reagent to study estrogen receptor signaling in biochemical and cellular assays. It is provided as a high-purity small molecule (CAS 1443752-77-9) with recommended solubility in DMSO and intended for research use only.
  • R-enantiomer isomer for use as a stereochemical control.
  • Selective estrogen receptor degrader activity suitable for ERα and ERβ studies.
  • High purity appropriate for biochemical and cellular assays.
  • Available in small pack sizes for experimental testing.
  • Soluble in DMSO for assay preparation.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-37393