Learn More
Medchemexpress LLC (R)-PF-04991532 | 1215197-37-7 | 99.4% | 396.36 | C18H19F3N4O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10018110MM 1ML
Encompass_Preferred
PF-04991532 is a hepatoselective glucokinase activator for preclinical research, showing potent activation with reported EC50 values of ~80 nM in human and ~100 nM in rat. Supplied as a 10 mM solution in DMSO, it is intended for laboratory research use.
  • Potent, hepatoselective glucokinase activator with low-nanomolar EC50s.
  • Ready-to-use 10 mM solution in DMSO for biochemical and cellular assays.
  • High chemical purity (99.4%) for consistent experimental results.
  • Molecular weight 396.36 and formula C18H19F3N4O3 for reference.
  • Stable when stored according to supplier recommendations to preserve activity.
Catalog No. 50-002-15327