Learn More
Medchemexpress LLC (R)-Phenylglycine tert-butyl ester hydrochloride | 256478-95-2 | MFCD03093480 | ≥97.0% | 243.73 g/mol | C12H18ClNO2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW042009A
Encompass_Preferred
(R)-Phenylglycine tert-butyl ester hydrochloride is the (R)-enantiomer of phenylglycine, protected as a tert-butyl ester and supplied as the hydrochloride salt for research use. It is a white to off-white solid with molecular weight 243.73 g/mol and reported purity ≥97.0%.
  • Chiral (R)-enantiomer for stereospecific synthesis.
  • Tert-butyl ester protection facilitates downstream deprotection.
  • Hydrochloride salt improves stability and handling as a solid.
  • High purity (≥97.0%) appropriate for research applications.
  • Available in multiple pack sizes, including 25 g.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years).
Catalog No. 50-002-09531