Learn More
Selleck Chemical LLC R-Phycoerythrin E2856-5MG
SDP
Supplier:  Selleck Chemical LLC E28565MG
Encompass_Preferred
R-Phycoerythrin(R-PE) phycobiliproteins are isolated from Heterosiphonia japonica R-Phycoerythrin is a novel strong fluorescent probe contains four chromophore-carrying subunits which exhibits extremely bright red-orange fluorescence
Catalog No. 50-000-39692