Learn More
Meso Scale Discovery R-PLEX Human CD30/TNFRSF8 Assay, K151M5R-2, Singleplex, 96-well plate, LLOD 190 pg/mL
SDP
Supplier:  Meso Scale Discovery K151M5R2
Encompass
The MESO SCALE DISCOVERY R-PLEX Human CD30/TNFRSF8 Assay is a singleplex immunoassay designed for the quantitative detection of human CD30/TNFRSF8. This assay leverages the R-PLEX platform, ensuring sensitive and precise measurements essential for biomarker research in oncology and cancer studies. Its compatibility with a variety of MSD instruments allows for versatile laboratory applications.
  • Provides sensitive and precise measurement of human CD30/TNFRSF8.
  • Includes essential components for straightforward singleplex assay development.
  • Designed for use with a range of MESO SECTOR and QuickPlex instruments.
  • Suitable for various research applications, particularly in oncology and cancer.
  • Lower limit of detection is 190 pg/mL.
  • Usage statement indicates for research use only.
  • Compatible with 96-well plate format.
  • Multi-component storage conditions required.
Catalog No. NC3073015