Learn More
Meso Scale Discovery R-PLEX Human AB42 6E10 ASYQP
SDP
Supplier:  Meso Scale Discovery K1518SR22
Encompass
The Meso Scale Discovery R-PLEX Human A 42/A 40 Assay Kit is a multiplex immunoassay for measuring A 42 and A 40 in human samples like CSF serum and plasma It uses electrochemiluminescence for high sensitivity and low sample volume The assay requires 50 L or less and runs in under 3 hours The kit includes a 96-well plate with antibodies detection antibodies read buffer calibrators controls diluent and seals It measures the A 42/A 40 ratio a key Alzheimer s biomarker
Catalog No. 50-000-80257