Learn More
Meso Scale Discovery R-PLEX Human AChE ASY
SDP
Supplier:  Meso Scale Discovery K151AHVR2
Encompass
The Meso Scale Discovery acetylcholinesterase assay kit measures acetylcholinesterase activity in samples like serum plasma cell lysates and tissue homogenates It uses electrochemiluminescence for high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate detection antibody buffers calibrator and substrate Detection limits range from 0 156 to 200 U/L with a total assay time of about 3 5 hours enabling reliable quantification in research
Catalog No. 50-000-79093