Learn More
Meso Scale Discovery R-PLEX Human AChE ASYQP
SDP
Supplier:  Meso Scale Discovery K151AHVR22
Encompass
The R-PLEX Human ACHE Assay Kit from Meso Scale Discovery is used to quantitatively measure acetylcholinesterase (ACHE) levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection combined with patterned arrays The assay has high sensitivity and a broad dynamic range It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and controls The total assay time is 3 5 hours This product can aid in understanding ACHE biology and the effects of drugs toxins and diseases
Catalog No. 50-000-81910