Learn More
Meso Scale Discovery R-PLEX Human ADAM 9 ASYQP
SDP
Supplier:  Meso Scale Discovery K151ABHR22
Encompass
The R-PLEX Human ADAM-9 Assay is a multiplex immunoassay kit that enables simultaneous quantification of ADAM-9 and other biomarkers in human samples such as serum plasma and cell culture supernatants This assay utilizes the quantitative electrochemiluminescent Meso Scale Discovery (MSD) platform and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies specific for each biomarker calibrators detection antibodies read buffer wash buffer and plate seals The assay takes approximately 3 5 hours to complete after a 2 hour incubation
Catalog No. 50-000-79572