Learn More
Meso Scale Discovery R-PLEX Human ADAM-TS 13 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AABR22
Encompass
The R-PLEX Human ADAM-TS13 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that quantitatively measures ADAM-TS13 antigen in human serum plasma and other biological samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls diluent wash buffer read buffer and plate seals This assay allows for the precise quantification of ADAM-TS13 levels which aids in the diagnosis of thrombotic thrombocytopenic purpura (TTP)
Catalog No. 50-000-79482