Learn More
Meso Scale Discovery R-PLEX Human AGP ASY
SDP
Supplier:  Meso Scale Discovery K151JRR2
Encompass
The R-PLEX Human AGP Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring alpha-1-acid glycoprotein (AGP) in human serum plasma and other fluids This 96-well electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive AGP quantification The kit includes a plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay offers a quick way to measure AGP an acute phase protein elevated in inflammation
Catalog No. 50-000-80795