Learn More
Meso Scale Discovery R-PLEX Human Ang-1 ASYQP
SDP
Supplier:  Meso Scale Discovery K151YQR22
Encompass
The R-PLEX Human Ang-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Angiopoietin-1 levels in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology allowing simultaneous quantitative measurement of Ang-1 and other biomarkers The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81818