Learn More
Meso Scale Discovery R-PLEX Human Ang-4 ASYQP
SDP
Supplier:  Meso Scale Discovery K151Y3R22
Encompass
The R-PLEX Human Ang-4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Angiopoietin-4 levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required to perform the assay Results are obtained in as little as 3 5 hours This assay allows for the accurate and reliable quantification of Ang-4 levels within a sample
Catalog No. 50-000-79660