Learn More
Meso Scale Discovery R-PLEX Human Ang-like 4 ASYQP
SDP
Supplier:  Meso Scale Discovery K151Z3R22
Encompass
The R-PLEX Human Ang-like 4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Ang-like 4 levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required for the assay The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81805