Learn More
Meso Scale Discovery R-PLEX Human BMP-9/GDF-2 Assay, Singleplex format, 96-well plate, Research use only, Human BMP-9/GDF-2, Bone Disorders application, Compatible with S 600MM, SQ 120MM, Q 60MM, SQ 120, S 600, SI 6000, SI 2400

Supplier: Meso Scale Discovery K1519TR2
The MESO SCALE DISCOVERY R-PLEX Human BMP-9/GDF-2 Assay provides essential components for the measurement of human BMP-9/GDF-2. This singleplex immunoassay leverages the R-PLEX platform, allowing researchers to create customized multiplex panels easily. Designed strictly for research applications, the assay ensures reliable and validated performance specifications across various sample types.
- Analyte: Human BMP-9/GDF-2
- Platform: R-PLEX
- Assay format: Singleplex (Can be multiplexed with R-PLEX Assay Designer)
- Product type: Assay Kit
- Species: Human
- Application: Bone disorders
- Plate type: 96-well
- Instrument compatibility: S 600MM, SQ 120MM, Q 60MM, SQ 120, S 600, SI 6000, SI 2400
- Research use only: Yes
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.