Learn More
Meso Scale Discovery R-PLEX Human BTLA ASYQP
SDP
Supplier:  Meso Scale Discovery K151ALNR22
Encompass
The R-PLEX Human BTLA Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring BTLA in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity a 5-log dynamic range and low sample volume This assay quantifies BTLA crucial for downregulating lymphocyte activation and promoting self-tolerance With a 4 5-hour protocol minimal hands-on time and included standard curve reagents it efficiently profiles the BTLA immunomodulatory pathway
Catalog No. 50-000-82014