Learn More
Meso Scale Discovery R-PLEX Human CA9 ASY
SDP
Supplier:  Meso Scale Discovery K151H4R2
Encompass
The R-Plex Human CA9 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures Carbonic Anhydrase 9 (CA9) in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be used to measure CA9 levels for cancer biomarker research and drug development This multiplex kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer and 10X Wash Buffer
Catalog No. 50-000-79273