Learn More
Meso Scale Discovery R-PLEX Human CA9 ASYQP
SDP
Supplier:  Meso Scale Discovery K151H4R22
Encompass
The R-PLEX Human CA9 Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring human Carbonic Anhydrase 9 (CA9) in serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible CA9 results It requires 50 L of sample and is read on MSD s QuickPlex SQ 120 The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls offering a robust tool for cancer and hypoxia research
Catalog No. 50-000-82339