Learn More
Meso Scale Discovery R-PLEX Human Calbindin ASY
SDP
Supplier:  Meso Scale Discovery K151YUR2
Encompass
The R-PLEX Human Calbindin Assay from Meso Scale Diagnostics is a multiplex immunoassay for measuring calbindin in human serum plasma urine and other fluids It uses electrochemiluminescence (ECL) detection for high sensitivity a wide dynamic range and low sample volume This assay quantifies calbindin a calcium-binding protein in the intestine and kidneys aiding in diagnosing diseases related to calbindin levels The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents
Catalog No. 50-000-79824