Learn More
Meso Scale Discovery R-PLEX Human CD14 ASYQP
SDP
Supplier:  Meso Scale Discovery K151T1R22
Encompass
The R-PLEX Human CD14 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures CD14 levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls This assay allows for the quantitative measurement of CD14 which plays a key role in innate immune response The kit has high performance and reliability for biomarker research and drug development applications
Catalog No. 50-000-78561