Learn More
Meso Scale Discovery R-PLEX Human Chemerin ASY
SDP
Supplier:  Meso Scale Discovery K151ACBR2
Encompass
The R-PLEX Human Chemerin Assay from Meso Scale Discovery is a multiplex immunoassay that measures chemerin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay allows for the quantitative measurement of chemerin which plays a role in immune response inflammation and metabolic disorders It requires 50 L or less of sample and can perform up to 500 tests across two plates The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker A blocker D and calibrator
Catalog No. 50-000-80326