Learn More
Meso Scale Discovery R-PLEX Human cIAP1 ASYQP
SDP
Supplier:  Meso Scale Discovery K1517VR22
Encompass
The R-PLEX Human CIAP1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the levels of Cellular Inhibitor of Apoptosis Protein 1 (CIAP1) in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrators and controls The assay can detect CIAP1 in the range of 19 5-20 000 pg/mL and requires only 25 L of sample
Catalog No. 50-000-78913