Learn More
Meso Scale Discovery R-PLEX Human cIAP2 ASY
SDP
Supplier:  Meso Scale Discovery K1517WR2
Encompass
The R-PLEX Human CIAP2 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures cellular inhibitor of apoptosis protein 2 (CIAP2) in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the quantitative measurement of CIAP2 which plays a role in inhibiting apoptosis and is involved in inflammatory signaling pathways The kit includes a 96-well plate coated with capture antibody detection antibody read buffer sample diluent calibrator diluent and a vial of calibrator
Catalog No. 50-000-78950