Learn More
Meso Scale Discovery R-PLEX Human COMP/TSP-5 ASYQP
SDP
Supplier:  Meso Scale Discovery K151U0R22
Encompass
The R-PLEX Human Comp TSP-5 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring 5 human Th17-related proteins (IL-17A IL-17F IL-22 IL-23 IL-33) in serum plasma and cell culture supernatants It uses MSD s MULTI-ARRAY technology and includes plates with capture antibodies detection antibodies read buffer calibrators and controls The assay is sensitive reproducible has a broad dynamic range and requires only 12 5 L sample per well enabling high-throughput analysis of Th17 biomarkers related to autoimmunity and inflammation
Catalog No. 50-000-78524