Learn More
Meso Scale Discovery R-PLEX Human Corin/ATC2 ASY
SDP
Supplier:  Meso Scale Discovery K1510BR2
Encompass
The R-PLEX Human Corin (ATC2) Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures the levels of Corin (ATC2) in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrators and controls This assay allows for the quantitative measurement of Corin (ATC2) levels with high precision and accuracy
Catalog No. 50-000-80281