Learn More
Meso Scale Discovery R-PLEX Human Corin/ATC2 ASYQP
SDP
Supplier:  Meso Scale Discovery K1510BR22
Encompass
The R-PLEX Human Corin (AT2) Assay Kit from Meso Scale Discovery is used to quantitatively measure human corin in serum plasma cell culture supernatants cell and tissue lysates It utilizes an electrochemiluminescent detection method and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L of sample per well and takes approximately 4 hours to complete It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocking solution matrix solution and calibrator This assay kit can be used for biomarker discovery and validation
Catalog No. 50-000-78832