Learn More
Meso Scale Discovery R-PLEX Human COX-2/PTGS2 ASY
SDP
Supplier:  Meso Scale Discovery K151AJJR2
Encompass
The R-Plex Human COX-2 (PTGS2) Assay is a multiplex immunoassay kit by Meso Scale Diagnostics that enables the simultaneous quantitative measurement of COX-2 protein in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the rapid reproducible analysis of COX-2 an inducible enzyme involved in inflammation and cancer With minimal hands-on time it serves as an efficient tool for biomarker research and drug development
Catalog No. 50-000-81629