Learn More
Meso Scale Discovery R-PLEX Human Cystatin B ASY
SDP
Supplier:  Meso Scale Discovery K151AAKR2
Encompass
The R-PLEX Human Cystatin B Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for measuring Cystatin B in human serum plasma (EDTA heparin) and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for a rapid ultra-sensitive and cost-effective measurement The assay detects from 0 04 ng/mL to 1500 ng/mL requires 50 L of sample and is read on MSD s SECTOR Imager instruments offering a robust tool for studying Cystatin B levels
Catalog No. 50-000-80387