Learn More
Meso Scale Discovery R-PLEX Human E-Cadherin ASYQP
SDP
Supplier:  Meso Scale Discovery K151YXR22
Encompass
The R-Plex Human E-Cadherin Assay from Meso Scale Discovery is a multiplex immunoassay that measures E-Cadherin levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements The assay can monitor E-Cadherin levels over time and across experimental conditions It includes a 96-well plate coated with capture antibodies detection antibodies and calibrators to generate a standard curve The R-Plex Human E-Cadherin Assay delivers rapid precise quantification of E-Cadherin
Catalog No. 50-000-79215