Learn More
Meso Scale Discovery R-PLEX Human EGF ASYQP
SDP
Supplier:  Meso Scale Discovery K1515FR22
Encompass
The R-PLEX Human EGF Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Epidermal Growth Factor (EGF) in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator The assay has a typical sensitivity of 0 23 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81688