Learn More
Meso Scale Discovery R-PLEX Human Elastase-2 ASYQP
SDP
Supplier:  Meso Scale Discovery K151ABKR22
Encompass
The R-PLEX Human Elastase 2 Assay from Meso Scale Discovery is a multiplex immunoassay panel that measures Elastase 2 levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can be used to monitor Elastase 2 levels as a biomarker and requires only 50 L of sample per well The panel allows simultaneous measurement of Elastase 2 along with other biomarkers in human samples
Catalog No. 50-000-79875