Learn More
Meso Scale Discovery R-PLEX Human GFAP ASYQP
SDP
Supplier:  Meso Scale Discovery K1511MR22
Encompass
The R-PLEX Human GFAP Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of glial fibrillary acidic protein (GFAP) in human serum plasma and other biological samples The assay uses MSD s proprietary SECTOR Imager 6000 for high sensitivity detection of GFAP a biomarker expressed by astrocytes in the central nervous system The kit includes a 96-well plate coated with anti-GFAP capture antibody detection antibody read buffer sample diluent calibrator and complete instructions The assay has a typical sensitivity of 0 046 ng/mL and an upper limit of quantification of 1500 ng/mL
Catalog No. 50-000-80207