Learn More
Meso Scale Discovery R-PLEX Human IA-2/R-PTP-N ASY
SDP
Supplier:  Meso Scale Discovery K151AKAR2
Encompass
The Meso Scale Discovery K151AKAR-2 is an immunoassay kit for the quantitative measurement of IA-2 RPTP-N in human serum plasma and other biological samples The kit uses electrochemiluminescence technology for high sensitivity and wide dynamic range It includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for sample dilution and washing The assay has a typical sensitivity of 0 05 ng/mL and range of 0 05-1000 ng/mL It requires 2 5 hours total assay time The kit provides components for one 96-well plate and can test 40 samples in duplicate
Catalog No. 50-000-78433