Learn More
Meso Scale Discovery R-PLEX Human ICAM-3 ASY
SDP
Supplier:  Meso Scale Discovery K151ZCR2
Encompass
The R-PLEX Human ICAM-3 Assay Kit from Meso Scale Discovery measures Intercellular Adhesion Molecule 3 (ICAM-3) in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours with a dynamic range of 16-10 000 pg/mL offering a simple reproducible method to measure soluble ICAM-3 levels
Catalog No. 50-000-79240